Assay Details
| Gene Symbol: LOC100284299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157194.1 | NP_001150666.1 | 503 | 70 | ||
| GenBank mRNA | BT039296.1 | - | 339 | 70 | ||
| BT061657.1 | - | 430 | 70 | |||
| BT067991.1 | - | 483 | 70 | |||
| EU967819.1 | - | 503 | 70 | |||
More Information
Set Membership: |
|

LOC100284299
Genomic Map