Assay Details
| Gene Symbol: LOC100284299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157194.1 | NP_001150666.1 | 824 | 99 | ||
| GenBank mRNA | BT039296.1 | - | 660 | 99 | ||
| BT067991.1 | - | 804 | 99 | |||
| EU967819.1 | - | 824 | 99 | |||
More Information
Set Membership: |
|

LOC100284299
Genomic Map