Assay Details
| Gene Symbol: LOC100284299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157194.1 | NP_001150666.1 | 904 | 96 | ||
| GenBank mRNA | BT039296.1 | - | 740 | 96 | ||
| BT061657.1 | - | 1587 | 96 | |||
| BT067991.1 | - | 884 | 96 | |||
| EU967819.1 | - | 904 | 96 | |||
More Information
Set Membership: |
|

LOC100284299
Genomic Map