Assay Details
Gene Symbol: LOC100284728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157623.1 | NP_001151095.1 | 1401 | 102 | ||
GenBank mRNA | EU969487.1 | - | 1401 | 102 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
