Assay Details
| Gene Symbol: LOC100285667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158558.1 | NP_001152030.1 | 546 | 88 | ||
| GenBank mRNA | EU973336.1 | - | 546 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100285667| Gene Symbol: LOC100285667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158558.1 | NP_001152030.1 | 546 | 88 | ||
| GenBank mRNA | EU973336.1 | - | 546 | 88 | ||