Assay Details
| Gene Symbol: LOC100285667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158558.1 | NP_001152030.1 | 668 | 66 | ||
| GenBank mRNA | EU973336.1 | - | 668 | 66 | ||
More Information
Set Membership: |
|

LOC100285667
Genomic Map