Assay Details
| Gene Symbol: LOC100285804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158694.1 | NP_001152166.1 | 452 | 96 | ||
| GenBank mRNA | EU974049.1 | - | 452 | 96 | ||
More Information
Set Membership: |
|

LOC100285804
Genomic Map