Assay Details
| Gene Symbol: LOC100286189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159077.1 | NP_001152549.1 | 2094 | 65 | ||
| GenBank mRNA | EU976053.1 | - | 2094 | 65 | ||
More Information
Set Membership: |
|

LOC100286189
Genomic Map