Assay Details
| Gene Symbol: myb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111616.1 | NP_001105086.1 | 2052 | 95 | ||
| GenBank mRNA | AF458962.1 | - | 2052 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

myb2| Gene Symbol: myb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111616.1 | NP_001105086.1 | 2052 | 95 | ||
| GenBank mRNA | AF458962.1 | - | 2052 | 95 | ||