Assay Details
| Gene Symbol: met1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111716.1 | NP_001105186.1 | 3579 | 80 | ||
| GenBank mRNA | AF229183.1 | - | 1340 | 80 | ||
| AY093415.1 | - | 3579 | 80 | |||
| AY109471.1 | - | 3563 | 80 | |||
More Information
Set Membership: |
|

met1
Genomic Map