Assay Details
Gene Symbol: LOC542127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111759.1 | NP_001105229.1 | 514 | 60 | ||
GenBank mRNA | AY303682.1 | - | 514 | 60 |
Gene Symbol: LOC100501069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT060851.1 | - | 353 | 60 | ||
BT061899.1 | - | 351 | 60 | |||
BT061986.1 | - | 625 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
