Assay Details
| Gene Symbol: cyc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111917.1 | NP_001105387.1 | 809 | 88 | ||
| GenBank mRNA | U50064.1 | - | 809 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cyc2| Gene Symbol: cyc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111917.1 | NP_001105387.1 | 809 | 88 | ||
| GenBank mRNA | U50064.1 | - | 809 | 88 | ||
Set Membership: |
|