Assay Details
| Gene Symbol: his1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111960.1 | NP_001105430.1 | 679 | 99 | ||
| GenBank mRNA | X57077.1 | - | 679 | 99 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

his1| Gene Symbol: his1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111960.1 | NP_001105430.1 | 679 | 99 | ||
| GenBank mRNA | X57077.1 | - | 679 | 99 | ||
Set Membership: |
|