Assay Details
| Gene Symbol: amyb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112026.1 | NP_001105496.1 | 926 | 65 | ||
| GenBank mRNA | AY111979.1 | - | 926 | 65 | ||
| Z25871.1 | - | 926 | 65 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

amyb5| Gene Symbol: amyb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112026.1 | NP_001105496.1 | 926 | 65 | ||
| GenBank mRNA | AY111979.1 | - | 926 | 65 | ||
| Z25871.1 | - | 926 | 65 | |||