Assay Details
| Gene Symbol: fmi1(pki) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112028.1 | NP_001105498.1 | 144 | 75 | ||
| GenBank mRNA | Z29643.1 | - | 144 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fmi1(pki)| Gene Symbol: fmi1(pki) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112028.1 | NP_001105498.1 | 144 | 75 | ||
| GenBank mRNA | Z29643.1 | - | 144 | 75 | ||
Set Membership: |
|