Assay Details
| Gene Symbol: pco139601b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112197.1 | NP_001105667.1 | 3749 | 89 | ||
| GenBank mRNA | AY186247.1 | - | 3749 | 89 | ||
More Information
Set Membership: |
|

pco139601b
Genomic Map