Assay Details
| Gene Symbol: Smh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112200.1 | NP_001105670.1 | 552 | 82 | ||
| GenBank mRNA | AY280632.1 | - | 552 | 82 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smh6| Gene Symbol: Smh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112200.1 | NP_001105670.1 | 552 | 82 | ||
| GenBank mRNA | AY280632.1 | - | 552 | 82 | ||
Set Membership: |
|