Assay Details
| Gene Symbol: gl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112420.1 | NP_001105890.1 | 800 | 85 | ||
| GenBank mRNA | U41466.1 | - | 800 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gl15| Gene Symbol: gl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112420.1 | NP_001105890.1 | 800 | 85 | ||
| GenBank mRNA | U41466.1 | - | 800 | 85 | ||
Set Membership: |
|