Assay Details
| Gene Symbol: mrpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112472.1 | NP_001105942.1 | 4322 | 82 | ||
| GenBank mRNA | AY186245.1 | - | 4322 | 82 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrpa1| Gene Symbol: mrpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112472.1 | NP_001105942.1 | 4322 | 82 | ||
| GenBank mRNA | AY186245.1 | - | 4322 | 82 | ||
Set Membership: |
|