Assay Details
| Gene Symbol: lox6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112506.1 | NP_001105976.1 | 920 | 100 | ||
| GenBank mRNA | BT063943.1 | - | 916 | 100 | ||
| BT067473.1 | - | 916 | 100 | |||
| BT085507.1 | - | 908 | 100 | |||
| DQ335764.1 | - | 920 | 100 | |||
More Information
Set Membership: |
|

lox6
Genomic Map