Assay Details
| Gene Symbol: AGPLS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112588.1 | NP_001106058.1 | 631 | 92 | ||
| GenBank mRNA | EF694838.1 | - | 631 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGPLS3| Gene Symbol: AGPLS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112588.1 | NP_001106058.1 | 631 | 92 | ||
| GenBank mRNA | EF694838.1 | - | 631 | 92 | ||
Set Membership: |
|