Assay Details
| Gene Symbol: LOC100280229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153159.1 | NP_001146631.1 | 2297 | 95 | ||
| GenBank mRNA | BT055818.1 | - | 2297 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100280229| Gene Symbol: LOC100280229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153159.1 | NP_001146631.1 | 2297 | 95 | ||
| GenBank mRNA | BT055818.1 | - | 2297 | 95 | ||