Assay Details
| Gene Symbol: LOC100280783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153705.1 | NP_001147177.1 | 243 | 80 | ||
| GenBank mRNA | EU953751.1 | - | 243 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100280783| Gene Symbol: LOC100280783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153705.1 | NP_001147177.1 | 243 | 80 | ||
| GenBank mRNA | EU953751.1 | - | 243 | 80 | ||