Assay Details
| Gene Symbol: LOC100280783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153705.1 | NP_001147177.1 | 372 | 55 | ||
| GenBank mRNA | EU953751.1 | - | 372 | 55 | ||
More Information
Set Membership: |
|

LOC100280783
Genomic Map