Assay Details
| Gene Symbol: LOC100282184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155096.1 | NP_001148568.1 | 705 | 77 | ||
| GenBank mRNA | BT068542.1 | - | 705 | 77 | ||
| EU959960.1 | - | 683 | 77 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100282184| Gene Symbol: LOC100282184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155096.1 | NP_001148568.1 | 705 | 77 | ||
| GenBank mRNA | BT068542.1 | - | 705 | 77 | ||
| EU959960.1 | - | 683 | 77 | |||