Assay Details
| Gene Symbol: LOC100284258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157153.1 | NP_001150625.1 | 1315 | 81 | ||
| GenBank mRNA | BT053866.1 | - | 1272 | 81 | ||
| EU967681.1 | - | 1315 | 81 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100284258| Gene Symbol: LOC100284258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157153.1 | NP_001150625.1 | 1315 | 81 | ||
| GenBank mRNA | BT053866.1 | - | 1272 | 81 | ||
| EU967681.1 | - | 1315 | 81 | |||