Assay Details
| Gene Symbol: LOC100284274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157169.1 | NP_001150641.1 | 646 | 68 | ||
| GenBank mRNA | EU967752.1 | - | 646 | 68 | ||
More Information
Set Membership: |
|

LOC100284274
Genomic Map