Assay Details
| Gene Symbol: LOC100284692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157587.1 | NP_001151059.1 | 674 | 98 | ||
| GenBank mRNA | EU969348.1 | - | 674 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100284692| Gene Symbol: LOC100284692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157587.1 | NP_001151059.1 | 674 | 98 | ||
| GenBank mRNA | EU969348.1 | - | 674 | 98 | ||