Assay Details
| Gene Symbol: LOC100284692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157587.1 | NP_001151059.1 | 718 | 80 | ||
| GenBank mRNA | EU969348.1 | - | 718 | 80 | ||
More Information
Set Membership: |
|

LOC100284692
Genomic Map