Assay Details
| Gene Symbol: LOC100285256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158150.1 | NP_001151622.1 | 417 | 78 | ||
| GenBank mRNA | BT060686.1 | - | 1328 | 78 | ||
| EU971432.1 | - | 417 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100285256| Gene Symbol: LOC100285256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158150.1 | NP_001151622.1 | 417 | 78 | ||
| GenBank mRNA | BT060686.1 | - | 1328 | 78 | ||
| EU971432.1 | - | 417 | 78 | |||