Assay Details
Gene Symbol: LOC100285688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158579.1 | NP_001152051.1 | 638 | 87 | ||
GenBank mRNA | EU973426.1 | - | 638 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
