Assay Details
| Gene Symbol: LOC100285799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158689.1 | NP_001152161.1 | 576 | 81 | ||
| GenBank mRNA | EU974027.1 | - | 576 | 81 | ||
More Information
Set Membership: |
|

LOC100285799
Genomic Map