Assay Details
| Gene Symbol: LOC100285939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158828.1 | NP_001152300.1 | 182 | 72 | ||
| GenBank mRNA | EU974774.1 | - | 182 | 72 | ||
More Information
Set Membership: |
|

LOC100285939
Genomic Map