Assay Details
| Gene Symbol: LOC100286076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158964.1 | NP_001152436.1 | 328 | 85 | ||
| GenBank mRNA | EU975492.1 | - | 328 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100286076| Gene Symbol: LOC100286076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158964.1 | NP_001152436.1 | 328 | 85 | ||
| GenBank mRNA | EU975492.1 | - | 328 | 85 | ||