Assay Details
| Gene Symbol: LOC100272703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147156.1 | NP_001140628.1 | 863 | 98 | ||
| GenBank mRNA | BT037461.1 | - | 818 | 98 | ||
| BT039199.1 | - | 863 | 98 | |||
| EU968063.1 | - | 850 | 98 | |||
More Information
Set Membership: |
|

LOC100272703
Genomic Map