Assay Details
| Gene Symbol: chb102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147851.1 | NP_001141323.1 | 1063 | 60 | ||
| GenBank mRNA | BT041066.1 | - | 1063 | 60 | ||
| EU965400.1 | - | 1090 | 60 | |||
More Information
Set Membership: |
|

chb102
Genomic Map