Assay Details
| Gene Symbol: LOC100282053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154966.1 | NP_001148438.1 | 822 | 65 | ||
| GenBank mRNA | BT087337.1 | - | 761 | 65 | ||
| EU959350.1 | - | 822 | 65 | |||
More Information
Set Membership: |
|

LOC100282053
Genomic Map