Assay Details
| Gene Symbol: LOC100282389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155300.1 | NP_001148772.1 | 786 | 94 | ||
| GenBank mRNA | EU960739.1 | - | 786 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100282389| Gene Symbol: LOC100282389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155300.1 | NP_001148772.1 | 786 | 94 | ||
| GenBank mRNA | EU960739.1 | - | 786 | 94 | ||