Assay Details
Gene Symbol: LOC100282425 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155336.1 | NP_001148808.1 | 706 | 59 | ||
GenBank mRNA | BT068780.1 | - | 628 | 59 | ||
EU960851.1 | - | 706 | 59 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
