Assay Details
| Gene Symbol: LOC100283077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155979.1 | NP_001149451.1 | 452 | 82 | ||
| GenBank mRNA | EU963375.1 | - | 452 | 82 | ||
More Information
Set Membership: |
|

LOC100283077
Genomic Map