Assay Details
| Gene Symbol: LOC100283475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156375.1 | NP_001149847.1 | 338 | 78 | ||
| GenBank mRNA | EU964862.1 | - | 338 | 78 | ||
More Information
Set Membership: |
|

LOC100283475
Genomic Map