Assay Details
Gene Symbol: LOC100284259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157154.1 | NP_001150626.1 | 780 | 70 | ||
GenBank mRNA | EU967684.1 | - | 780 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
