Assay Details
| Gene Symbol: LOC100284546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157441.1 | NP_001150913.1 | 215 | 65 | ||
| GenBank mRNA | EU968811.1 | - | 215 | 65 | ||
More Information
Set Membership: |
|

LOC100284546
Genomic Map