Assay Details
| Gene Symbol: aap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159178.1 | NP_001152650.1 | 885 | 75 | ||
| GenBank mRNA | BT085955.1 | - | 852 | 75 | ||
| EU976635.1 | - | 885 | 75 | |||
More Information
Set Membership: |
|

aap1
Genomic Map