Assay Details
Gene Symbol: pco118508(516) | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146969.1 | NP_001140441.1 | 414 | 51 | ||
GenBank mRNA | BT038841.1 | - | 414 | 51 | ||
EU962942.1 | - | 426 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() |
