Assay Details
| Gene Symbol: csu525(rpL17) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001150266.1 | NP_001143738.1 | 940 | 109 | ||
| GenBank mRNA | EU962731.1 | - | 940 | 109 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

csu525(rpL17)| Gene Symbol: csu525(rpL17) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001150266.1 | NP_001143738.1 | 940 | 109 | ||
| GenBank mRNA | EU962731.1 | - | 940 | 109 | ||