Assay Details
| Gene Symbol: csu425(gct) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152318.1 | NP_001145790.1 | 1547 | 68 | ||
| GenBank mRNA | BT053985.1 | - | 1547 | 68 | ||
More Information
Set Membership: |
|

csu425(gct)
Genomic Map