Assay Details
| Gene Symbol: LOC100279375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152391.1 | NP_001145863.1 | 845 | 86 | ||
| GenBank mRNA | BT041580.1 | - | 758 | 86 | ||
| BT054137.1 | - | 845 | 86 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100279375| Gene Symbol: LOC100279375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152391.1 | NP_001145863.1 | 845 | 86 | ||
| GenBank mRNA | BT041580.1 | - | 758 | 86 | ||
| BT054137.1 | - | 845 | 86 | |||