Assay Details
| Gene Symbol: LOC100280076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153016.1 | NP_001146488.1 | 736 | 75 | ||
| GenBank mRNA | BT055522.1 | - | 736 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100280076| Gene Symbol: LOC100280076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153016.1 | NP_001146488.1 | 736 | 75 | ||
| GenBank mRNA | BT055522.1 | - | 736 | 75 | ||