Assay Details
Gene Symbol: LOC100280502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153422.1 | NP_001146894.1 | 1449 | 92 | ||
GenBank mRNA | BT065394.1 | - | 1449 | 92 | ||
EU952152.1 | - | 1356 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
